Trichloroacetic acid, 99+%, ACS reagent
Trichloroacetic acid, 99+%, ACS reagent
Trichloroacetic acid, 99+%, ACS reagent
Trichloroacetic acid, 99+%, ACS reagent
Thermo Scientific Chemicals
Trichloroacetic acid, 99+%, ACS reagent
CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass Bottle
Catalog number FSA421455000
Price (MYR)
775.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
775.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrichloroacetic acid
Name Note99+%
CAS76-03-9
Health Hazard 1Danger