p-Toluenesulfonic acid, pure, 12% in acetic acid
p-Toluenesulfonic acid, pure, 12% in acetic acid
p-Toluenesulfonic acid, pure, 12% in acetic acid
p-Toluenesulfonic acid, pure, 12% in acetic acid
Thermo Scientific Chemicals
p-Toluenesulfonic acid, pure, 12% in acetic acid
CAS: 104-15-4 | C7H8O3S | 172.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number FSA421222500
Price (MYR)
834.00
BOT
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
834.00
BOT
Add to cart
Chemical Identifiers
CAS104-15-4, 64-19-7
Specifications
Chemical Name or Materialp-Toluenesulfonic acid
ColorColorless to Yellow
Name Note12% in Acetic Acid
CAS104-15-4
Boiling Point116.0°C