N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis
CAS: 110-18-9 | C6H16N2 | 116.21 g/mol
Have Questions?
Change view
Quantity:
500 g
5 g
50 g
100 g
Packaging:
Glass bottle
Catalog number FSA420585000
Price (MYR)
2,006.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,006.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N,N',N'-Tetramethylethylenediamine
Name Note99%
CAS110-18-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger