Tetramethylammonium Hydroxide, 10% in Water
Tetramethylammonium Hydroxide, 10% in Water
Tetramethylammonium Hydroxide, 10% in Water
Tetramethylammonium Hydroxide, 10% in Water
Thermo Scientific Chemicals
Tetramethylammonium Hydroxide, 10% in Water
CAS: 75-59-2 | C4H13NO | 91.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA420515000
Price (MYR)
728.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
728.00
BOT
Add to cart
Chemical Identifiers
CAS75-59-2, 7732-18-5
Specifications
Chemical Name or MaterialTetramethylammonium hydroxide
Name Note10% in Water
CAS75-59-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
View more
Phase transfer catalyst
RUO – Research Use Only