Tetraethylammonium Hydroxide, 10% in Water
Tetraethylammonium Hydroxide, 10% in Water
Tetraethylammonium Hydroxide, 10% in Water
Tetraethylammonium Hydroxide, 10% in Water
Thermo Scientific Chemicals
Tetraethylammonium Hydroxide, 10% in Water
CAS: 77-98-5 | C8H21NO | 147.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA420281000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS77-98-5, 7732-18-5
Specifications
Chemical Name or MaterialTetraethylammonium hydroxide
ColorColorless
Name Note10% in Water
CAS77-98-5
Boiling Point102.0°C
View more
Phase transfer catalyst
RUO – Research Use Only