Tellurium, 99+%, (trace metal basis), powder
Tellurium, 99+%, (trace metal basis), powder
Tellurium, 99+%, (trace metal basis), powder
Tellurium, 99+%, (trace metal basis), powder
Thermo Scientific Chemicals
Tellurium, 99+%, (trace metal basis), powder
CAS: 13494-80-9 | Te | 127.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA420020100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point990.0°C
Chemical Name or MaterialTellurium
Melting Point450.0°C
Name Note99+%
CAS13494-80-9