Salicylic acid, 99+%, ACS reagent
Salicylic acid, 99+%, ACS reagent
Salicylic acid, 99+%, ACS reagent
Salicylic acid, 99+%, ACS reagent
Thermo Scientific Chemicals
Salicylic acid, 99+%, ACS reagent
CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA419225000
Price (MYR)
536.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
536.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSalicylic acid
CAS69-72-7
Melting Point158.0°C to 161.0°C
Boiling Point211.0°C (20.0 mmHg)
Health Hazard 1SIGNAL WORD: DANGER!
View more
Biological buffer
RUO – Research Use Only