Rhodamine 6G, 99%, pure
Rhodamine 6G, 99%, pure
Rhodamine 6G, 99%, pure
Rhodamine 6G, 99%, pure
Thermo Scientific Chemicals
Rhodamine 6G, 99%, pure
CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA419011000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodamine 6G
Melting Point290.0°C
Name Note99%
CAS989-38-8
Health Hazard 2GHS H Statement:
Harmful if swallowed.
View more
Indicator
RUO – Research Use Only