1,10-Phenanthroline monohydrate, ACS reagent
1,10-Phenanthroline monohydrate, ACS reagent
1,10-Phenanthroline monohydrate, ACS reagent
1,10-Phenanthroline monohydrate, ACS reagent
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, ACS reagent
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA417121000
Price (MYR)
2,350.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,350.00
EA
Add to cart
Specifications
Chemical Name or Material1, 10-Phenanthroline monohydrate
CAS5144-89-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
CAS66-71-7
View more
Indicator
RUO – Research Use Only