L(+)-Penicillamine, 98%
L(+)-Penicillamine, 98%
L(+)-Penicillamine, 98%
L(+)-Penicillamine, 98%
Thermo Scientific Chemicals
L(+)-Penicillamine, 98%
CAS: 1113-41-3 | C5H11NO2S | 149.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA416870050
Price (MYR)
1,520.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,520.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL(+)-Penicillamine
Name Note98%
CAS1113-41-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement