Pectin, pract.
Thermo Scientific Chemicals
Pectin, pract.
CAS: 9000-69-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA416862500
Price (MYR)
705.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
705.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPectin
ColorOrange to Orange-Red
pH3.0 to 3.8 (1% soln.)
CAS9000-69-5
Physical FormPowder