Pararosaniline chloride, 96%, pure
Pararosaniline chloride, 96%, pure
Pararosaniline chloride, 96%, pure
Pararosaniline chloride, 96%, pure
Thermo Scientific Chemicals
Pararosaniline chloride, 96%, pure
CAS: 569-61-9 | C19H17N3·HCl | 323.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA416801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPararosaniline Chloride
Melting Point268.0°C to 270.0°C
CAS569-61-9
ColorGreen
Health Hazard 1GHS Signal Word: Danger
View more
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA
RUO – Research Use Only