Palmitic acid, sodium salt, 98%
Palmitic acid, sodium salt, 98%
Palmitic acid, sodium salt, 98%
Palmitic acid, sodium salt, 98%
Thermo Scientific Chemicals
Palmitic acid, sodium salt, 98%
CAS: 408-35-5 | C16H31NaO2 | 278.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA416700050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPalmitic acid, sodium salt
CAS408-35-5
ColorWhite
EINECS Number206-988-1
Loss on Drying2% max. (110°C, 4 hrs)