Octyl isocyanate, 99%
Octyl isocyanate, 99%
Octyl isocyanate, 99%
Octyl isocyanate, 99%
Thermo Scientific Chemicals
Octyl isocyanate, 99%
CAS: 3158-26-7 | C9H17NO | 155.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number FSA416460100
Price (MYR)
637.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
637.00
BOT
Add to cart
Specifications
Chemical Name or MaterialOctyl isocyanate
Name Note99%
CAS3158-26-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger