1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
Thermo Scientific Chemicals
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
CAS: 521-24-4 | C10H5NaO5S | 260.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA415400250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Naphthoquinone-4-sulfonic acid sodium salt
Name Note99%
CAS521-24-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning