Methyl valerate, 99%
Methyl valerate, 99%
Methyl valerate, 99%
Methyl valerate, 99%
Thermo Scientific Chemicals
Methyl valerate, 99%
CAS: 624-24-8 | C6H12O2 | 116.16 g/mol
Have Questions?
Catalog NumberQuantity
FSA41505025025 g
Catalog number FSA415050250
Price (MYR)
1,019.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,019.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl valerate
Name Note99%
Acidity1mg KOH/g max.
CAS624-24-8
Health Hazard 1GHS Signal Word: Warning