Methyl palmitate, 95%
Methyl palmitate, 95%
Methyl palmitate, 95%
Methyl palmitate, 95%
Thermo Scientific Chemicals
Methyl palmitate, 95%
CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA414685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl palmitate
Name Note95%
CAS112-39-0
Melting Point28.0°C