Methyl oleate, technical
Methyl oleate, technical
Methyl oleate, technical
Methyl oleate, technical
Thermo Scientific Chemicals
Methyl oleate, technical
CAS: 67762-38-3 | C19H36O2 | 296.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA414645000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl oleate
CAS67762-38-3
Melting Point-20.0°C
ColorYellow to Amber
Recommended StorageRefrigerator +4°C