Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm
Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm
Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm
Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm
Thermo Scientific Chemicals
Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm
CAS: 7439-95-4 | Mg | 24.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA413381000
Price (MYR)
1,635.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,635.00
EA
Add to cart
Specifications
Chemical Name or MaterialMagnesium
ColorWhite
Name Note99+%
CAS7439-95-4
Melting Point651.0°C