Lauric acid, sodium salt, 98%
Lauric acid, sodium salt, 98%
Lauric acid, sodium salt, 98%
Lauric acid, sodium salt, 98%
Thermo Scientific Chemicals
Lauric acid, sodium salt, 98%
CAS: 629-25-4 | C12H23NaO2 | 222.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA413040250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLauric acid, sodium salt
Name Note99%
CAS629-25-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.