3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™
Thermo Scientific Chemicals
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™
CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA412721000
Price (MYR)
351.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
351.00
EA
Add to cart
Specifications
Chemical Name or Material3-Methyl-1-butanol
CAS123-51-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin