Iron(0) pentacarbonyl
Iron(0) pentacarbonyl
Iron(0) pentacarbonyl
Iron(0) pentacarbonyl
Thermo Scientific Chemicals
Iron(0) pentacarbonyl
CAS: 13463-40-6 | C5FeO5 | 195.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA412601000
Price (MYR)
630.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
630.00
EA
Add to cart
Specifications
Chemical Name or MaterialIron pentacarbonyl
Name Note99.5%
CAS13463-40-6
ColorBrown to Orange-Yellow
Health Hazard 1Danger