IR-125, pure, laser grade
IR-125, pure, laser grade
IR-125, pure, laser grade
IR-125, pure, laser grade
Thermo Scientific Chemicals
IR-125, pure, laser grade
CAS: 3599-32-4 | C43H47N2NaO6S2 | 774.967 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA412541000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIR-125
Melting Point235.0°C
CAS3599-32-4
Health Hazard 1HYGROSCOPIC
Health Hazard 2GHS H Statement: