Hydroxylamine hydrochloride, ACS reagent
Hydroxylamine hydrochloride, ACS reagent
Hydroxylamine hydrochloride, ACS reagent
Hydroxylamine hydrochloride, ACS reagent
Thermo Scientific Chemicals
Hydroxylamine hydrochloride, ACS reagent
CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA412055000
Price (MYR)
953.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
953.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydroxylamine hydrochloride
Melting Point155°C to 158°C
Name Note96+%
CAS5470-11-1
ColorWhite
View more
Reduction reagent
RUO – Research Use Only