Hydriodic acid, ACS reagent, 55% in water, without stabilizer
Hydriodic acid, ACS reagent, 55% in water, without stabilizer
Thermo Scientific Chemicals
Hydriodic acid, ACS reagent, 55% in water, without stabilizer
CAS: 10034-85-2 | HI | 127.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Catalog number FSA411840500
Price (MYR)
464.00
EA
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (MYR)
464.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydriodic acid
CAS10034-85-2
Boiling Point127.0°C
Health Hazard 1GHS Signal Word: Danger
Density1.7000 g/mL