L-Histidine hydrochloride monohydrate, 98%
L-Histidine hydrochloride monohydrate, 98%
L-Histidine hydrochloride monohydrate, 98%
L-Histidine hydrochloride monohydrate, 98%
Thermo Scientific Chemicals
L-Histidine hydrochloride monohydrate, 98%
CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA411735000
Price (MYR)
2,066.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
2,066.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Histidine hydrochloride monohydrate
Name Note98%
Ammonium200ppm max.
CAS5934-29-2
pH3 to 5 (2.5% soln. at 25°C)