Hexadecyltrimethylammonium Chloride, 97%
Hexadecyltrimethylammonium Chloride, 97%
Hexadecyltrimethylammonium Chloride, 97%
Hexadecyltrimethylammonium Chloride, 97%
Thermo Scientific Chemicals
Hexadecyltrimethylammonium Chloride, 97%
CAS: 112-02-7 | C19H42ClN | 320.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA411410050
Price (MYR)
761.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
761.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexadecyltrimethylammonium chloride
CAS112-02-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
View more
Phase transfer catalyst
RUO – Research Use Only