Heptafluorobutyric acid, 99%, for protein sequence analysis
Heptafluorobutyric acid, 99%, for protein sequence analysis
Heptafluorobutyric acid, 99%, for protein sequence analysis
Heptafluorobutyric acid, 99%, for protein sequence analysis
Heptafluorobutyric acid, 99%, for protein sequence analysis
CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass Bottle
Catalog number FSA411221000
Price (MYR)
-
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHeptafluorobutyric Acid
CAS375-22-4
Absorbance(0.1% aqueous solution), (0.1% aqueous solution) at 0nm, 0.004 max. at 280nm, 0.006 max. at 254nm, 0.15 max. at 230nm
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement