Glyoxylic acid, 50% in water
Glyoxylic acid, 50% in water
Glyoxylic acid, 50% in water
Glyoxylic acid, 50% in water
Thermo Scientific Chemicals
Glyoxylic acid, 50% in water
CAS: 298-12-4 | C2H2O3 | 74.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number FSA411065000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGlyoxylic acid
ColorColorless to Yellow
Name Note50% in water
CAS298-12-4
Boiling Point111.0°C