D(+)-Glucose, ACS Reagent, anhydrous
D(+)-Glucose, ACS Reagent, anhydrous
D(+)-Glucose, ACS Reagent, anhydrous
D(+)-Glucose, ACS Reagent, anhydrous
Thermo Scientific Chemicals
D(+)-Glucose, ACS Reagent, anhydrous
CAS: 50-99-7 | C6H12O6 | 180.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Catalog number FSA410955000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Glucose
Name NoteAnhydrous, For analysis ACS
CAS50-99-7
Melting Point146.0°C