Formic acid, 88%, ACS reagent
Formic acid, 88%, ACS reagent
Formic acid, 88%, ACS reagent
Formic acid, 88%, ACS reagent
Thermo Scientific Chemicals
Formic acid, 88%, ACS reagent
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
Catalog number FSA410775000
Price (MYR)
462.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
462.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFormic acid
CAS64-18-6
Melting Point8.0°C
Boiling Point101.0°C
Health Hazard 1Danger