Fast Green FCF, pure, certified
Fast Green FCF, pure, certified
Fast Green FCF, pure, certified
Fast Green FCF, pure, certified
Thermo Scientific Chemicals
Fast Green FCF, pure, certified
CAS: 2353-45-9 | C37H34N2Na2O10S3 | 808.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA410531000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFast Green FCF
Melting Point290.0°C
CAS2353-45-9
Health Hazard 1Warning
Absorption Ratio0.98 to 1.2