2-Ethylhexyl acrylate, 99+%, stabilized
2-Ethylhexyl acrylate, 99+%, stabilized
2-Ethylhexyl acrylate, 99+%, stabilized
2-Ethylhexyl acrylate, 99+%, stabilized
Thermo Scientific Chemicals
2-Ethylhexyl acrylate, 99+%, stabilized
CAS: 103-11-7 | C11H20O2 | 184.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA410110250
Price (MYR)
300.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
300.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Ethylhexyl acrylate
Acidity0.01% max. (as acrylic acid)
CAS103-11-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning