Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Thermo Scientific Chemicals
Dipropyl phthalate, 99+%
CAS: 131-16-8 | C14H18O4 | 250.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA409161000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDipropyl phthalate
CAS131-16-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Suspected of causing cancer.
Health Hazard 3GHS P Statement:
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.