3,5-Dinitrobenzoyl Chloride, 99%
3,5-Dinitrobenzoyl Chloride, 99%
3,5-Dinitrobenzoyl Chloride, 99%
3,5-Dinitrobenzoyl Chloride, 99%
Thermo Scientific Chemicals
3,5-Dinitrobenzoyl Chloride, 99%
CAS: 99-33-2 | C7H3ClN2O5 | 230.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA408711000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 5-Dinitrobenzoyl chloride
CAS99-33-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0