N,N-Dimethyl-p-phenylenediamine, 97%
N,N-Dimethyl-p-phenylenediamine, 97%
N,N-Dimethyl-p-phenylenediamine, 97%
N,N-Dimethyl-p-phenylenediamine, 97%
Thermo Scientific Chemicals
N,N-Dimethyl-p-phenylenediamine, 97%
CAS: 99-98-9 | C8H12N2 | 136.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA408461000
Price (MYR)
708.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
708.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethyl-p-phenylenediamine
Name NotePractical
CAS99-98-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger