Dimethylglyoxime, 99+%, ACS reagent
Dimethylglyoxime, 99+%, ACS reagent
Dimethylglyoxime, 99+%, ACS reagent
Dimethylglyoxime, 99+%, ACS reagent
Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent
CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA408335000
Price (MYR)
1,126.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,126.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethylglyoxime
CAS95-45-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.
Toxic if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediate