Specifications
Chemical Name or MaterialDimethylamine
CAS124-40-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Highly flammable liquid and vapor.
CAS7732-18-5
View more