N,N-Dimethylallylamine, 98%
N,N-Dimethylallylamine, 98%
N,N-Dimethylallylamine, 98%
N,N-Dimethylallylamine, 98%
Thermo Scientific Chemicals
N,N-Dimethylallylamine, 98%
CAS: 2155-94-4 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA407980250
Price (MYR)
1,323.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,323.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylallylamine
Name Note98%
CAS2155-94-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger