1,3-Dibromo-2-propanol, 95%
1,3-Dibromo-2-propanol, 95%
1,3-Dibromo-2-propanol, 95%
1,3-Dibromo-2-propanol, 95%
Thermo Scientific Chemicals
1,3-Dibromo-2-propanol, 95%
CAS: 96-21-9 | C3H6Br2O | 217.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA406541000
Price (MYR)
1,319.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,319.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 3-Dibromo-2-propanol
Name Notepract., 85%
CAS96-21-9
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning