trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
Thermo Scientific Chemicals
trans-1,4-Dibromo-2-butene, 97%
CAS: 821-06-7 | C4H6Br2 | 213.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA406461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-1, 4-Dibromo-2-butene
Name Note97%
CAS821-06-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.