Dibenzyldithiocarbamic acid zinc salt, 95%
Dibenzyldithiocarbamic acid zinc salt, 95%
Dibenzyldithiocarbamic acid zinc salt, 95%
Dibenzyldithiocarbamic acid zinc salt, 95%
Thermo Scientific Chemicals
Dibenzyldithiocarbamic acid zinc salt, 95%
CAS: 14726-36-4 | C30H28N2S4Zn | 610.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA406440250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDibenzyldithiocarbamic acid zinc salt
CAS14726-36-4
Health Hazard 1GHS Signal Word: Warning
Melting Point183.0°C to 188.0°C