Dextrin
Dextrin
Dextrin
Thermo Scientific Chemicals
Dextrin
CAS: 9004-53-9 | C18H32O16 | 504.438 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA406285000
Price (MYR)
578.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
578.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDextrin
ColorWhite to Yellow
CAS9004-53-9
Physical FormCrystalline Powder
RTECS NumberHH9450000