Chromium(VI) oxide, 98+%, ACS reagent
Chromium(VI) oxide, 98+%, ACS reagent
Chromium(VI) oxide, 98+%, ACS reagent
Chromium(VI) oxide, 98+%, ACS reagent
Thermo Scientific Chemicals
Chromium(VI) oxide, 98+%, ACS reagent
CAS: 1333-82-0 | CrO3 | 99.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA405235000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChromium(VI) oxide
Melting Point196.0°C
Name Note98+%
CAS1333-82-0
ColorBrown-Red to Purple
View more
Oxidation reagent
RUO – Research Use Only