Activated charcoal, DARCO™, 4-12 mesh, decolorizing
Activated charcoal, DARCO™, 4-12 mesh, decolorizing
Activated charcoal, DARCO™, 4-12 mesh, decolorizing
Activated charcoal, DARCO™, 4-12 mesh, decolorizing
Thermo Scientific Chemicals
Activated charcoal, DARCO™, 4-12 mesh, decolorizing
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
3 kg
Packaging:
View all
Glass bottle
Catalog number FSA403995000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialActivated charcoal,DARCO*
Name NoteDARCO™, 4 to 12 Mesh, Decolorizing
CAS7440-44-0
Melting Point3652.0°C
ColorBlack
View more
Filter aid
RUO – Research Use Only