tert-Butyl Ethyl Ether, 97%
tert-Butyl Ethyl Ether, 97%
tert-Butyl Ethyl Ether, 97%
tert-Butyl Ethyl Ether, 97%
Thermo Scientific Chemicals
tert-Butyl Ethyl Ether, 97%
CAS: 637-92-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA403530250
Price (MYR)
1,154.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,154.00
BOT
Add to cart
Specifications
Chemical Name or Materialtert-Butyl ethyl ether
CAS637-92-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
May cause drowsiness or dizziness.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.