2-Bromoethyl methyl ether, 95%
2-Bromoethyl methyl ether, 95%
2-Bromoethyl methyl ether, 95%
2-Bromoethyl methyl ether, 95%
Thermo Scientific Chemicals
2-Bromoethyl methyl ether, 95%
CAS: 6482-24-2 | C3H7BrO | 138.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA402981000
Price (MYR)
1,653.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,653.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromoethyl methyl ether
CAS6482-24-2
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0