4-Bromoaniline, 96%, pract.
4-Bromoaniline, 96%, pract.
4-Bromoaniline, 96%, pract.
4-Bromoaniline, 96%, pract.
Thermo Scientific Chemicals
4-Bromoaniline, 96%, pract.
CAS: 106-40-1 | C6H6BrN | 172.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA402871000
Price (MYR)
447.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
447.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Bromoaniline
Name Note(pract), 96%
CAS106-40-1
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger