Benzyldimethylhexadecylammonium chloride hydrate, 97%
Benzyldimethylhexadecylammonium chloride hydrate, 97%
Benzyldimethylhexadecylammonium chloride hydrate, 97%
Benzyldimethylhexadecylammonium chloride hydrate, 97%
Thermo Scientific Chemicals
Benzyldimethylhexadecylammonium chloride hydrate, 97%
CAS: 122-18-9 | C25H46ClN | 396.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA402070050
Price (MYR)
203.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
203.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyldimethylhexadecylammonium chloride hydrate
Name Note97 - 101%
CAS122-18-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
View more
Phase transfer catalyst
RUO – Research Use Only