4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
Thermo Scientific Chemicals
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
CAS: 116-63-2 | C10H9NO4S | 239.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA400851000
Price (MYR)
1,252.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,252.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Amino-3-hydroxy-1-naphthalenesulfonic acid
Name Note98+%
CAS116-63-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: